Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Piperidinecarbonitrile, 97%, Thermo Scientific™

Catalog No. AC200240500
Change view
Click to view available options
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC200240500 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC200240500 Supplier Thermo Scientific Chemicals Supplier No. 200240500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1530-87-6
CAS Min % 96.0
CAS Max % 100.0
Density 0.9500g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H10N2
Refractive Index 1.4690 to 1.4710 (20°C, 589nm)
Quantity 50g
MDL Number MFCD00006477
Beilstein 20, 56
Synonym 1-piperidinecarbonitrile, 1-cyanopiperidine, piperidinocarbonitrile, n-cyanopiperidine, pentamethylenecyanamide, cyanopiperidine, cyano-piperidine, piperidine nitrile, piperidinecarbonitrile, 1-cyanopiperidine #
InChI Key NVPICXQHSYQKGM-UHFFFAOYSA-N
SMILES N#CN1CCCCC1
IUPAC Name piperidine-1-carbonitrile
Molecular Weight (g/mol) 110.16
PubChem CID 73736
Formula Weight 110.16
Percent Purity ≥96.0%
Chemical Name or Material 1-Piperidinecarbonitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-233-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.