Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-piperidinecarboxylic acid, 2,2-dimethyl-5-oxo-, 1,1-dimethylethyl ester | 1894533-96-0 | MFCD31718688 | 96.8% | 227.30 | C12H21NO3 | 250 MG
SDP

Catalog No. 5000281498
Encompass_Preferred

tert-Butyl 2,2-dimethyl-5-oxopiperidine-1-carboxylate is a Boc-protected piperidine derivative used as a synthetic intermediate in medicinal chemistry and drug discovery. The solid is provided with analytical verification (1H NMR, LCMS) and a Certificate of Analysis reporting LCMS purity of 96.84%. Molecular formula C12H21NO3 and molecular weight 227.30 are supplied for formulation and calculation purposes.

  • Boc-protected piperidine intermediate suitable for organic synthesis.
  • High purity by LCMS (96.8%).
  • Supplied as a white to off-white solid.
  • Analytical data provided: 1H NMR and LCMS.
  • Storage instructions included for solid and solutions.
  • Molecular weight 227.30 and formula C12H21NO3.

Catalog No. 50-002-81498 Supplier Medchemexpress LLC Supplier No. HYW451756250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl 2,2-dimethyl-5-oxopiperidine-1-carboxylate is a Boc-protected piperidine derivative used as a synthetic intermediate in medicinal chemistry and drug discovery. The solid is provided with analytical verification (1H NMR, LCMS) and a Certificate of Analysis reporting LCMS purity of 96.84%. Molecular formula C12H21NO3 and molecular weight 227.30 are supplied for formulation and calculation purposes.

  • Boc-protected piperidine intermediate suitable for organic synthesis.
  • High purity by LCMS (96.8%).
  • Supplied as a white to off-white solid.
  • Analytical data provided: 1H NMR and LCMS.
  • Storage instructions included for solid and solutions.
  • Molecular weight 227.30 and formula C12H21NO3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.