Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Piperidineethanol 98.0+%, TCI America™
SDP

Catalog No. P034825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P034825G 25 g
P0348500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P034825G Supplier TCI America Supplier No. P034825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3040-44-6
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00006512
InChI Key KZTWONRVIPPDKH-UHFFFAOYSA-N
Synonym 2-piperidinoethanol, 1-piperidineethanol, 2-piperidin-1-yl ethanol, 1-2-hydroxyethyl piperidine, n-2-hydroxyethyl piperidine, n-piperidineethanol, 2-1-piperidinyl ethanol, n-hydroxyethyl piperidine, beta-piperidylethanol, beta-piperidinoethanol
PubChem CID 18232
ChEBI CHEBI:61238
IUPAC Name 2-piperidin-1-ylethanol
SMILES C1CCN(CC1)CCO

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 g
Formula Weight 129.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Piperidineethanol
RTECSNumber TM8052000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.