Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Piperonylpiperazine, 97%, Thermo Scientific™

Catalog No. AAL1562514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1562514 25 g
AAL1562506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1562514 Supplier Thermo Scientific Chemicals Supplier No. L1562514

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32231-06-4
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD00005834
InChI Key NBOOZXVYXHATOW-UHFFFAOYSA-N
Synonym 1-piperonylpiperazine, 1-3,4-methylenedioxybenzyl piperazine, 1-benzo d 1,3 dioxol-5-ylmethyl piperazine, 1-1,3-benzodioxol-5-ylmethyl piperazine, 1-1,3-benzodioxol-5-yl-methyl piperazine, 1-benzo 1,3 dioxol-5-ylmethyl-piperazine, piperazine, 1-1,3-benzodioxol-5-ylmethyl, 1-3,4-methylendioxybenzyl piperazine, piperazine, 1-3,4-methylenedioxybenzyl, 5-piperazinylmethyl-2h-benzo d 1,3-dioxolene
PubChem CID 94426
IUPAC Name 1-(1,3-benzodioxol-5-ylmethyl)piperazine
SMILES C1CN(CCN1)CC2=CC3=C(C=C2)OCO3

Specifications

Melting Point 40°C to 45°C
Boiling Point 147°C to 149°C (2 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3259
Beilstein 885038
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 1-Piperonylpiperazine
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 250-968-5
RTECSNumber TM1550000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.