Learn More
Medchemexpress LLC 1-piperoylpiperidine-d10 | 94-62-2 | MFCD00005839 | 295.40 g/mol | C17H9D10NO3 | 2.5 MG
SDP
Catalog No. 5000284156
Encompass_Preferred
Piperin-d10 is a deuterium-labeled analogue of piperine supplied for analytical and research use. It serves as a mass-shifted internal standard or tracer for mass spectrometry and metabolic studies, enabling improved quantitation and tracking of piperine-related chemistry. Supplied in a small pack appropriate for method development and analytical use.
  • Deuterium-labeled internal standard for MS quantitation.
  • Mass-shifted analogue of piperine for tracer studies.
  • Suitable for analytical method development and validation.
  • Small pack size for research-scale experiments.
  • Certificate of analysis and reports available upon request.
Catalog No. 50-002-84156 Supplier Medchemexpress LLC Supplier No. HYW6543202.5MG
Piperin-d10 is a deuterium-labeled analogue of piperine supplied for analytical and research use. It serves as a mass-shifted internal standard or tracer for mass spectrometry and metabolic studies, enabling improved quantitation and tracking of piperine-related chemistry. Supplied in a small pack appropriate for method development and analytical use.
  • Deuterium-labeled internal standard for MS quantitation.
  • Mass-shifted analogue of piperine for tracer studies.
  • Suitable for analytical method development and validation.
  • Small pack size for research-scale experiments.
  • Certificate of analysis and reports available upon request.