Learn More
Heathrow Scientific 1-Place Cardboard Slide Holders, 25/pk.
Description
- Color: Natural
- Material: Cardboard
- Material: Cardboard
- Quantity: Pack of 25
Specifications
Specifications
| Type | Slide Mailer |
| Slide Capacity | 1 |
| Color | Natural |
| Dimensions (L x W x H) | 3.7 x 1.7 x 0.2 in. (95 x 42 x 5 mm) |
| Material | Cardboard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.