Learn More
Bio Rad Laboratories 1% TAE Wide Mini ReadyAgarose Precast Gel, 15.6 x 10 cm, 32-well, with ethidium bromide
Catalog No. NC9838004
Encompass
32-well precast wide mini gel, TAE, 1.0% plus ethidium bromide, multichannel pipet compatible, 15.6 x 10 cm (tray size)
Catalog No. NC9838004 Supplier Bio Rad Laboratories Supplier No. 1613050
32-well precast wide mini gel, TAE, 1.0% plus ethidium bromide, multichannel pipet compatible, 15.6 x 10 cm (tray size)