Learn More
THORLABS INC Ø1/2" Post Holder, Spring-Loaded Hex-Locking Thumbscrew, L = 1" | 5 Pack
Catalog No. NC1553746
Encompass
hese standard post holders are designed with a wide, square relief cut that provides two lines of contact for highly stable post mounting. A spring-loaded plunger within the aluminum thumbscrew holds a steel post firmly within the post holder prior to final tightening. This design allows one-handed positioning of post-mounted devices. Thorlabs' imperial and metric post holders are equipped with our 3/16" (TS25H) and 5 mm (TS6H/M) spring-loaded hex-locking thumbscrews, respectively (also available separately below). These thumbscrews should not be tightened beyond the maximum torque of 28 in·lbs (3.2 N·m).
Catalog No. NC1553746 Supplier THORLABS INC Supplier No. PH1P5
hese standard post holders are designed with a wide, square relief cut that provides two lines of contact for highly stable post mounting. A spring-loaded plunger within the aluminum thumbscrew holds a steel post firmly within the post holder prior to final tightening. This design allows one-handed positioning of post-mounted devices. Thorlabs' imperial and metric post holders are equipped with our 3/16" (TS25H) and 5 mm (TS6H/M) spring-loaded hex-locking thumbscrews, respectively (also available separately below). These thumbscrews should not be tightened beyond the maximum torque of 28 in·lbs (3.2 N·m).