Learn More
Medchemexpress LLC 1-Propanamine, 3-[4-[6-[3-(dimethylamino)propoxy]-2-benzoxazolyl]phenoxy]-N,N-dimethyl- | 1219962-49-8 | 98.8% | 397.51 | C23H31N3O3 | 50MG
SDP
Catalog No. 5000235110
Encompass_Preferred
AT791 is a small-molecule, orally bioavailable inhibitor of Toll-like receptors 7 and 9 (TLR7, TLR9). It inhibits TLR7- and TLR9-mediated signaling in multiple human and mouse cell types and blocks DNA-TLR9 interaction in vitro, making it useful for preclinical studies of innate immune signaling and inflammation. Chemical identifiers include CAS 1219962-49-8, formula C23H31N3O3, and molecular weight 397.51 g/mol.
  • Potent TLR7 and TLR9 inhibition in human and mouse cells.
  • Orally bioavailable small molecule suitable for in vivo dosing studies.
  • Demonstrated blockade of DNA-TLR9 interaction in vitro.
  • Available in multiple purified pack sizes for research use.
  • High purity reported in batch documentation (>98%).
Catalog No. 50-002-35110 Supplier Medchemexpress LLC Supplier No. HY12460350MG
AT791 is a small-molecule, orally bioavailable inhibitor of Toll-like receptors 7 and 9 (TLR7, TLR9). It inhibits TLR7- and TLR9-mediated signaling in multiple human and mouse cell types and blocks DNA-TLR9 interaction in vitro, making it useful for preclinical studies of innate immune signaling and inflammation. Chemical identifiers include CAS 1219962-49-8, formula C23H31N3O3, and molecular weight 397.51 g/mol.
  • Potent TLR7 and TLR9 inhibition in human and mouse cells.
  • Orally bioavailable small molecule suitable for in vivo dosing studies.
  • Demonstrated blockade of DNA-TLR9 interaction in vitro.
  • Available in multiple purified pack sizes for research use.
  • High purity reported in batch documentation (>98%).