Learn More
Medchemexpress LLC 1-Propanamine, 3-(triethoxysilyl)- | 919-30-2 | MFCD00008207 | 99.7% | 10 G
SDP
Catalog No. 5000410005
Encompass_Preferred
3-Aminopropyltriethoxysilane (APTES) acts as a strong adhesive for immobilizing biomolecules, such as antibodies and enzymes, onto silicon and its derivatives. It functions as a spacer to increase the spatial freedom of biomolecules, enhancing specific activity and creating a more stable, sensitive, and highly biocompatible bioanalytical platform on various solid materials, electrode materials, nanomaterials, and nanocomposites.
  • Functions as strong adhesive for biomolecule immobilization.
  • Increases spatial freedom and specific activity of biomolecules.
  • Creates stable, sensitive, and biocompatible bioanalytical platforms.
  • Exhibits hydrolytic instability in aqueous solutions.
  • Ethoxy group hydrolyzes into ethanol and trienol group.
  • Half-lives are 56 hours at 10°C and 8.4 hours at 24°C at neutral pH.
Catalog No. 50-004-10005 Supplier Medchemexpress LLC Supplier No. HYD017510G
3-Aminopropyltriethoxysilane (APTES) acts as a strong adhesive for immobilizing biomolecules, such as antibodies and enzymes, onto silicon and its derivatives. It functions as a spacer to increase the spatial freedom of biomolecules, enhancing specific activity and creating a more stable, sensitive, and highly biocompatible bioanalytical platform on various solid materials, electrode materials, nanomaterials, and nanocomposites.
  • Functions as strong adhesive for biomolecule immobilization.
  • Increases spatial freedom and specific activity of biomolecules.
  • Creates stable, sensitive, and biocompatible bioanalytical platforms.
  • Exhibits hydrolytic instability in aqueous solutions.
  • Ethoxy group hydrolyzes into ethanol and trienol group.
  • Half-lives are 56 hours at 10°C and 8.4 hours at 24°C at neutral pH.