Learn More
Medchemexpress LLC 1-Propanamine, N,N-dimethyl-2,3-bis[(9Z,12Z)-9,12-octadecadien-1-yloxy]- | 871258-12-7 | 99.5% | 616.06 | 250 MG
SDP
Catalog No. 5000439908
Encompass_Preferred
DLinDMA is an ionizable cationic lipid that serves as a key lipid component and benchmark for stable nucleic acid lipid particles (SNALPs). It is utilized for siRNA delivery. The structure of DLinDMA consists of three primary regions: the hydrocarbon chains, the linker, and the headgroup. It has demonstrated virtually indistinguishable blood pharmacokinetic profiles in mice.
  • Ionizable cationic lipid
  • Key lipid component of stable nucleic acid lipid particles (SNALPs)
  • Used for siRNA delivery
Catalog No. 50-004-39908 Supplier Medchemexpress LLC Supplier No. 250MGHY112757
DLinDMA is an ionizable cationic lipid that serves as a key lipid component and benchmark for stable nucleic acid lipid particles (SNALPs). It is utilized for siRNA delivery. The structure of DLinDMA consists of three primary regions: the hydrocarbon chains, the linker, and the headgroup. It has demonstrated virtually indistinguishable blood pharmacokinetic profiles in mice.
  • Ionizable cationic lipid
  • Key lipid component of stable nucleic acid lipid particles (SNALPs)
  • Used for siRNA delivery