Learn More
Medchemexpress LLC 1-propanaminium, 3-carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxohexyl)oxy]-, chloride | 2483831-95-2 | 99.0% | 298.82 g/mol | C13H23D3ClNO4 | 5 MG
SDP
Catalog No. 5000264232
Encompass_Preferred
L-Hexanoylcarnitine-d3 (chloride) is the deuterium-labeled analogue of L-hexanoylcarnitine provided as the chloride salt for use as an isotopically labeled internal standard in mass spectrometry and analytical workflows. It is supplied as a high-purity, isotopically enriched reference material for quantitative assays.
Catalog No. 50-002-64232 Supplier Medchemexpress LLC Supplier No. HY113144AS5MG
L-Hexanoylcarnitine-d3 (chloride) is the deuterium-labeled analogue of L-hexanoylcarnitine provided as the chloride salt for use as an isotopically labeled internal standard in mass spectrometry and analytical workflows. It is supplied as a high-purity, isotopically enriched reference material for quantitative assays.