Learn More
Medchemexpress LLC 1-propanaminium, 3-carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-, chloride | 18828-58-5 | MFCD00209823 | 98.1% | 253.72 g·mol⁻¹ | C10H20ClNO4 | 5 MG
SDP
Catalog No. 5000211438
Encompass_Preferred
Propionyl-DL-carnitine chloride is the racemic propionyl ester of carnitine presented as the chloride salt. It is provided as a solid research reagent for biochemical and cellular studies, including inflammation-related investigations, and is offered in small, precisely measured quantities with documented purity.
  • Carnitine derivative useful for inflammation research.
  • Chloride salt in solid form for stable handling and storage.
  • High purity specified at 98.1% for reproducible results.
  • Molecular formula C10H20ClNO4; molecular weight 253.72 g·mol⁻¹.
  • Supplied in small milligram packages suitable for analytical and in vitro work.
Catalog No. 50-002-11438 Supplier Medchemexpress LLC Supplier No. HY1010185MG
Propionyl-DL-carnitine chloride is the racemic propionyl ester of carnitine presented as the chloride salt. It is provided as a solid research reagent for biochemical and cellular studies, including inflammation-related investigations, and is offered in small, precisely measured quantities with documented purity.
  • Carnitine derivative useful for inflammation research.
  • Chloride salt in solid form for stable handling and storage.
  • High purity specified at 98.1% for reproducible results.
  • Molecular formula C10H20ClNO4; molecular weight 253.72 g·mol⁻¹.
  • Supplied in small milligram packages suitable for analytical and in vitro work.