Learn More
Medchemexpress LLC 1-propanesulfonamide derivative (PROTAC BRAF-V600E degrader-1) | 2417296-84-3 | 98.0% | C48H54F2N10O10S | 10MG
SDP
Catalog No. 5000259384
Encompass_Preferred
PROTAC BRAF-V600E degrader-1 is a research-grade targeted protein degrader that selectively induces proteasomal degradation of the oncogenic BRAF-V600E mutant. It shows low-nanomolar affinity for BRAF variants and is intended for cellular and biochemical research applications.
  • Selective degradation of BRAF-V600E via the ubiquitin-proteasome system.
  • Low-nanomolar binding affinity to BRAF and BRAF-V600E for potent activity.
  • High purity suitable for cellular assays and biochemical studies.
  • Available in small-mass sizes convenient for research workflows.
  • Provided for research use only; not for clinical applications.
Catalog No. 50-002-59384 Supplier Medchemexpress LLC Supplier No. HY13748710MG
PROTAC BRAF-V600E degrader-1 is a research-grade targeted protein degrader that selectively induces proteasomal degradation of the oncogenic BRAF-V600E mutant. It shows low-nanomolar affinity for BRAF variants and is intended for cellular and biochemical research applications.
  • Selective degradation of BRAF-V600E via the ubiquitin-proteasome system.
  • Low-nanomolar binding affinity to BRAF and BRAF-V600E for potent activity.
  • High purity suitable for cellular assays and biochemical studies.
  • Available in small-mass sizes convenient for research workflows.
  • Provided for research use only; not for clinical applications.