Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Propanesulfonic acid, 2-hydroxy-3-[[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]amino]- | 68399-81-5 | 98.0% | 25 G
SDP

Catalog No. 5000414468
Encompass_Preferred

TAPSO is a common biological buffer and pH stabilization reagent. It has an effective pH ranging from 7-8 and pKa values ranging from 7.5-9.0. This compound contains Tris groups and exhibits reactive activity with zwitterionic glycine peptides.

  • Common biological buffer
  • pH stabilization reagent
  • Effective pH ranging from 7-8
  • pKa values ranging from 7.5-9.0
  • Contains Tris groups
  • Reactive with zwitterionic glycine peptides
  • Suitable for research applications

Catalog No. 50-004-14468 Supplier Medchemexpress LLC Supplier No. 25GHYD0878
May include imposed supplier surcharges.
Add to Cart
Add to Cart

TAPSO is a common biological buffer and pH stabilization reagent. It has an effective pH ranging from 7-8 and pKa values ranging from 7.5-9.0. This compound contains Tris groups and exhibits reactive activity with zwitterionic glycine peptides.

  • Common biological buffer
  • pH stabilization reagent
  • Effective pH ranging from 7-8
  • pKa values ranging from 7.5-9.0
  • Contains Tris groups
  • Reactive with zwitterionic glycine peptides
  • Suitable for research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.