Learn More
Medchemexpress LLC 1-Propanesulfonic acid, 3-(bis(2-hydroxyethyl)amino)-2-hydroxy- | 68399-80-4 | MFCD00038353 | 98.0% | 100 G
SDP
Catalog No. 5000410411
Encompass_Preferred
DIPSO is a biological zwitterionic buffer with a useful pH range of 7.0 to 8.2. It can interfere with meiotic regulation in mouse oocytes and exhibits surfactant activity at 10 mM. This product is for research use only.
  • Useful pH range of 7.0 to 8.2
  • Can interfere with meiotic regulation in mouse oocytes
  • Exhibits surfactant activity at 10 mM
Catalog No. 50-004-10411 Supplier Medchemexpress LLC Supplier No. HYD0872100G
DIPSO is a biological zwitterionic buffer with a useful pH range of 7.0 to 8.2. It can interfere with meiotic regulation in mouse oocytes and exhibits surfactant activity at 10 mM. This product is for research use only.
  • Useful pH range of 7.0 to 8.2
  • Can interfere with meiotic regulation in mouse oocytes
  • Exhibits surfactant activity at 10 mM