Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Propanesulfonic acid, 3-(cyclohexylamino)-2-hydroxy- | 73463-39-5 | 98.0% | 25 G
SDP

Catalog No. 5000413782
Encompass_Preferred

CAPSO is a biological zwitterionic buffer with a useful pH range from 8.9 to 10.3. It is intended for research use only.

  • Molecular weight: 237.32
  • Formula: C9H19NO4S
  • Appearance: solid
  • Color: white to off-white
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility: DMSO: 25 mg/mL (105.34 mM)

Catalog No. 50-004-13782 Supplier Medchemexpress LLC Supplier No. 25GHYD0870
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAPSO is a biological zwitterionic buffer with a useful pH range from 8.9 to 10.3. It is intended for research use only.

  • Molecular weight: 237.32
  • Formula: C9H19NO4S
  • Appearance: solid
  • Color: white to off-white
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility: DMSO: 25 mg/mL (105.34 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.