Learn More
Sigma Aldrich Fine Chemicals Biosciences 1-Propanol suitable for HPLC, >=99.9% | 71-23-8 | MFCD00002941 | 1L
Catalog No. NC2171808
Encompass
1-Propanol suitable for HPLC, >=99.9% | Purity: >=99.9% | Mol Wt: 60.1 | 71-23-8 | MFCD00002941 | 1L
This item is not returnable. View return policy
Catalog No. NC2171808 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348711L
1-Propanol suitable for HPLC, >=99.9% | Purity: >=99.9% | Mol Wt: 60.1 | 71-23-8 | MFCD00002941 | 1L