Learn More
U.S. Pharmacopeia 1-Propanol, 71-23-8, MFCD00002941, 3 AMP
Catalog No. NC0940411
Encompass
Linear Formula: CH3CH2CH2OH, Molecular Weight: 60.10, bp: 97 °C, mp: −127 °C, Synonym: Propyl alcohol.
Catalog No. NC0940411 Supplier U.S. Pharmacopeia Supplier No. 1570406
Linear Formula: CH3CH2CH2OH, Molecular Weight: 60.10, bp: 97 °C, mp: −127 °C, Synonym: Propyl alcohol.