Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, 99+%, Extra Pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass Bottle
Metal drum
71-23-8
C3H8O
60.096
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
71-23-8
C3H8O
60.096
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
CAS Min % 99.0
CAS Max % 100.0
Additional Information Vapor Pressure: 25mbar at 20°C
Assay Percent Range 99+%
Linear Formula CH3CH2CH2OH
Beilstein 01, IV, 1413
Merck Index 15, 7955
Formula Weight 60.1
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Density 0.8000g/mL
Infrared Spectrum Authentic
Packaging Metal drum
Refractive Index 1.3840 to 1.3860
Residue after Evaporation 0.005% max.
Solubility Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
Specific Gravity 0.8
Water 0.5% max.
Flash Point 15°C
Viscosity 2.2 mPa.s (20°C)
Boiling Point 97.0°C
Color Undesignated
Melting Point -127.0°C
Quantity 25L
Chemical Name or Material 1-Propanol
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger