Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, 99+%

Catalog No. AAA199020F
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA199020F 2500 mL
AAA19902AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA199020F Supplier Thermo Scientific Chemicals Supplier No. A199020F
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 71-23-8 | C3H8O | 60.10 g/mol

1-Propanol is used as a solvent and an entrainer in azeotropic distillation. It is used as a precursor to prepare propyl acetate, propylamide and propyl halides by reacting with acetic acid, ammonia and halogens respectively. It is actively used as a solvent in inks for flexographic printing, which is a technique used for printing. It is added to cleaners, floor polishes and metal degreasing fluids and is used as an additional solvent in the manufacture of adhesives. It enhances the drying characteristics of alkyd resins, electrodeposition paints and baking finishes in coating industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Propanol is used as a solvent and an entrainer in azeotropic distillation. It is used as a precursor to prepare propyl acetate, propylamide and propyl halides by reacting with acetic acid, ammonia and halogens respectively. It is actively used as a solvent in inks for flexographic printing, which is a technique used for printing. It is added to cleaners, floor polishes and metal degreasing fluids and is used as an additional solvent in the manufacture of adhesives. It enhances the drying characteristics of alkyd resins, electrodeposition paints and baking finishes in coating industry.

Solubility
Miscible with water, alcohols, ketones, aldehydes, ethers, glycols, aromatic and aliphatic hydrocarbons.

Notes
Highly flammable. Incompatible with alkali metals, alkaline earths, aluminum, oxidizing agents, nitro compounds.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127°C
Density 0.804
Quantity 2500 mL
Flash Point 15°C (59°F)
Odor Alcohol-like
Refractive Index 1.385
UN Number UN1274
Beilstein 1098242
Merck Index 14,7842
Boiling Point 97°C
Solubility Information Miscible with water,alcohols,ketones,aldehydes,ethers,glycols,aromatic and aliphatic hydrocarbons.
Formula Weight 60.1
Percent Purity ≥99%
Chemical Name or Material 1-Propanol
Show More Show Less
Hazard Category H225-H318-H336
Hazard Statement GHS H Statement
H225-H318-H336
Highly flammable liquid and vapour.
Causes serious eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: N-PROPANOL
EINECSNumber 200-746-9
RTECSNumber UH8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.