Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, Analytical Standard, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193428 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8788 5 mL
11-101-8349 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8788 Supplier MilliporeSigma Supelco Supplier No. 965665MLF
Only null left
Add to Cart
Add to Cart

Specifications

CAS 71-23-8
Vapor Pressure 10 mmHg (147°C)
Vapor Density 2.1 (vs. air)
Density 0.804 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3CH2CH2OH
MDL Number MFCD00002941
Melting Point -127°C
Boiling Point 97°C
Synonym Propyl alcohol
Quantity 5 mL
Formula Weight 60.10
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.