Learn More
Sigma Aldrich Fine Chemicals Biosciences 1 Propanol suitable for HP1L
Catalog No. 502908237
Encompass
1-Propanol is a linear alcohol. It exhibits Debye-type relaxation process which ressembles the -relaxation and Johari-Goldstein type relaxation features of the supercooled liquids (nonhydrogen-bonding type). It has been tested as a substitute of the fuel for various fuel cells. Its biosynthesis from glucose using Escherichia coli strain has been proposed.
Catalog No. 50-290-8237 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348716X1L
Add to Cart
Add to Cart
1-Propanol is a linear alcohol. It exhibits Debye-type relaxation process which ressembles the -relaxation and Johari-Goldstein type relaxation features of the supercooled liquids (nonhydrogen-bonding type). It has been tested as a substitute of the fuel for various fuel cells. Its biosynthesis from glucose using Escherichia coli strain has been proposed.