Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, ACS reagent

Catalog No. AC445775000
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445775000 500 mL
AC445770010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445775000 Supplier Thermo Scientific Chemicals Supplier No. 445775000
  • $85.00
    $67.65 / Each of 1

    Save $17.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 71-23-8 | C3H8O | 60.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127.0°C
Density 0.8000g/mL
Quantity 500 mL
Flash Point 15°C
Refractive Index 1.3840 to 1.3860
Specific Gravity 0.8
Linear Formula CH3CH2CH2OH
Viscosity 2.2 mPa.s (20°C)
Water 0.2% max.
Boiling Point 97.0°C
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
Formula Weight 60.1
Percent Purity ≥99.5% (GC) 0.01% max. (Ethanol) (GC) 0.01% max. (Methanol) (GC) 0.05% max. (Isopropyl alcohol) (GC)
Grade ACS
Chemical Name or Material 1-Propanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement May cause drowsiness or dizziness. Causes serious eye damage. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.