Learn More
Medchemexpress LLC 1-Pyrenamin 250mg
SDP
Catalog No. 5000205737
Encompass_Preferred
1-Pyrenamin (1-Pyrenamine purified by sublimation) is utilized to create highly efficient and stable catalyst supports in direct methanol fuel cells through noncovalent functionalization of graphite nanosheets[1]
Catalog No. 50-002-05737 Supplier Medchemexpress LLC Supplier No. 250MGHYW094751
1-Pyrenamin (1-Pyrenamine purified by sublimation) is utilized to create highly efficient and stable catalyst supports in direct methanol fuel cells through noncovalent functionalization of graphite nanosheets[1]