Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pyreneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P16255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1625-5G 5 g
P1625-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1625-5G Supplier TCI America Supplier No. P16255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 164461-18-1
Melting Point 251°C
Molecular Formula C16H11BO2
Quantity 5 g
Synonym 1-pyrenylboronic acid, 1-pyreneboronic acid, pyrene-1-boronic acid, 1-boronopyrene, 1-pyrenyl boronic acid
InChI Key MWEKPLLMFXIZOC-UHFFFAOYSA-N
SMILES B(C1=C2C=CC3=CC=CC4=C3C2=C(C=C1)C=C4)(O)O
IUPAC Name pyren-1-ylboronic acid
Molecular Weight (g/mol) 246.072
PubChem CID 5084102
Formula Weight 246.07
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.