Learn More
Medchemexpress LLC 1-Pyrenebutanoic acid, 2,5-dioxo-1-pyrrolidinyl ester | 114932-60-4 | 99.9% | 385.41 | 500 MG
SDP
Catalog No. 5000454257
Encompass_Preferred
1-Pyrenebutyric acid N-hydroxysuccinimide ester (PANHS) is a linker and interface coupling agent utilized in the preparation of electrochemical biosensors. It is frequently employed as an activation reagent for carboxylic acids in organic chemistry or biochemistry.
  • Attaches antibodies onto CNT-FET circuits.
  • Used as an interfacial coupling agent to immobilize SARS-CoV-2 spike antibodies on graphene sheets for field-effect transistor-based biosensor devices to detect SARS-CoV-2.
Catalog No. 50-004-54257 Supplier Medchemexpress LLC Supplier No. HYW013151500MG
1-Pyrenebutyric acid N-hydroxysuccinimide ester (PANHS) is a linker and interface coupling agent utilized in the preparation of electrochemical biosensors. It is frequently employed as an activation reagent for carboxylic acids in organic chemistry or biochemistry.
  • Attaches antibodies onto CNT-FET circuits.
  • Used as an interfacial coupling agent to immobilize SARS-CoV-2 spike antibodies on graphene sheets for field-effect transistor-based biosensor devices to detect SARS-CoV-2.