Learn More
Invitrogen™ 1-pyrenebutanoic acid, succinimidyl ester
Description
The amine-reactive 1-pyrenebutanoic acid, succinimidyl ester can be used to create environment-sensitive bioconjugates with this unique fluorophore.
Specifications
Specifications
| Quantity | 100 mg |
| Product Type | 1-Pyrenebutanoic Acid Succinimidyl Ester |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | Pyrene |
| Label Type | Classic Dyes |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.