Learn More
Apexbio Technology LLC 1-Pyrenebutyric acid 3443-45-6 250mg
SDP
Catalog No. 5000095095
Encompass_Preferred
1-Pyrenebutyric acid (CAS 3443-45-6) is a fluorescent probe used in fluorescence-based DNA assays and biosensing applications This compound contains a polyaromatic pyrene moiety enabling stable noncovalent interactions with biomolecules and - stacking-mediated formation of organized monolayers on graphene and related substrates 1-Pyrenebutyric acid facilitates biomolecule immobilization and bioconjugation by serving as a linker in the functionalization of nanostructured materials It exhibits fluorescence sensitivity to molecular binding events supporting the detection of nucleic acids Based on these properties 1-Pyrenebutyric acid is utilized in fluorescence biosensing surface functionalization and the study of biomolecular interactions
Catalog No. 50-000-95095 Supplier Apexbio Technology LLC Supplier No. A4717250
1-Pyrenebutyric acid (CAS 3443-45-6) is a fluorescent probe used in fluorescence-based DNA assays and biosensing applications This compound contains a polyaromatic pyrene moiety enabling stable noncovalent interactions with biomolecules and - stacking-mediated formation of organized monolayers on graphene and related substrates 1-Pyrenebutyric acid facilitates biomolecule immobilization and bioconjugation by serving as a linker in the functionalization of nanostructured materials It exhibits fluorescence sensitivity to molecular binding events supporting the detection of nucleic acids Based on these properties 1-Pyrenebutyric acid is utilized in fluorescence biosensing surface functionalization and the study of biomolecular interactions