Learn More
Medchemexpress LLC 1-Pyrenebutyric acid 5g
SDP
Catalog No. 5000204785
Encompass_Preferred
1-Pyrenebutyric acid is a fluorescence probe whose fluorescence lifetime depends on local oxygen and free radical concentrations 1-Pyrenebutyric acid is used in fluorescence determination of DNA 1-Pyrenebutyric acid can be used as a linker for biomolecules to form a self-assembled monolayer on grapheme 1-Pyrenebutyric acid can also be used for the measurement of free radicals in solution and in living cells[1][2][3][4]
Catalog No. 50-002-04785 Supplier Medchemexpress LLC Supplier No. HYW1030475G
1-Pyrenebutyric acid is a fluorescence probe whose fluorescence lifetime depends on local oxygen and free radical concentrations 1-Pyrenebutyric acid is used in fluorescence determination of DNA 1-Pyrenebutyric acid can be used as a linker for biomolecules to form a self-assembled monolayer on grapheme 1-Pyrenebutyric acid can also be used for the measurement of free radicals in solution and in living cells[1][2][3][4]