Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pyrenebutyric Acid N-hydroxysuccinimide Ester, Sigma-Aldrich

Catalog No. 01258830 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-830 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-830 Supplier MilliporeSigma Supelco Supplier No. 4570781G
Add to Cart
Add to Cart

1-Pyrenebutyric acid N-hydroxysuccinimide ester has been used for NAD+ tethering on multi-walled carbon nanotubes (MWCNTs). It has been used in the preparation of laccase cathode.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114932-60-4
Molecular Weight (g/mol) 385.41 g/mol

Specifications

Quantity 1 g
Percent Purity 95%
Physical Form Crystalline Powder, Crystals or Chunks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.