Learn More
Medchemexpress LLC 1-Pyrenedecanoic acid | 64701-47-9 | MFCD00056577 | 99.3% | 372.50 g/mol | C26H28O2 | 10mg
SDP
Catalog No. 5000269749
Encompass_Preferred
1-Pyrenedecanoic acid is a biochemical reagent and can be used for membrane fluidity assessment[1]
Catalog No. 50-002-69749 Supplier Medchemexpress LLC Supplier No. HYW71392610MG
1-Pyrenedecanoic acid is a biochemical reagent and can be used for membrane fluidity assessment[1]