Learn More
Medchemexpress LLC 1-Pyrenemethanol | 24463-15-8 | 99.7% | 232.28 | 1 G
Supplier: Medchemexpress LLC HYW0096411G
1-Pyrenemethanol is an organic compound used to prepare graphene films with excellent thermal conductivity, flexibility, and mechanical properties. It binds to graphene oxide sheets through hydrogen bonds and π-π interactions to form GO-PyM films, enhancing the dispersion and stability of the material.
- Used to prepare graphene films with excellent thermal conductivity, flexibility, and mechanical properties
- Binds to graphene oxide sheets through hydrogen bonds and π-π interactions to form GO-PyM films
- Enhances the dispersion and stability of the material