Learn More
Medchemexpress LLC 1-Pyrenemethanol | 24463-15-8 | 99.7% | 232.28 | 5 G
SDP
Catalog No. 5000366912
Encompass_Preferred
1-Pyrenemethanol is an organic compound that can be used to prepare graphene films with excellent thermal conductivity, flexibility, and mechanical properties. It binds to graphene oxide sheets through hydrogen bonds and π-π interactions to form GO-PyM films, which increases the dispersion and stability of the material.
  • Prepares graphene films
  • Has excellent thermal conductivity
  • Has excellent flexibility
  • Has excellent mechanical properties
  • Binds to graphene oxide sheets through hydrogen bonds and π-π interactions
  • Increases material dispersion
  • Increases material stability
Catalog No. 50-003-66912 Supplier Medchemexpress LLC Supplier No. HYW0096415G
1-Pyrenemethanol is an organic compound that can be used to prepare graphene films with excellent thermal conductivity, flexibility, and mechanical properties. It binds to graphene oxide sheets through hydrogen bonds and π-π interactions to form GO-PyM films, which increases the dispersion and stability of the material.
  • Prepares graphene films
  • Has excellent thermal conductivity
  • Has excellent flexibility
  • Has excellent mechanical properties
  • Binds to graphene oxide sheets through hydrogen bonds and π-π interactions
  • Increases material dispersion
  • Increases material stability