Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-pyrenesulfonic acid sodium | 59323-54-5 | MFCD00003710 | 99.4% | 304.30 g/mol | C16H9NaO3S | 250 MG
SDP

Catalog No. 5000282656
Encompass_Preferred

1-Pyrenesulfonic acid sodium is the sodium salt of 1-pyrenesulfonic acid, a water-soluble, fluorescent aromatic sulfonate supplied as a solid research reagent. It is used as a fluorescent probe and analytical standard in life-science and chemical research; typical listed purity is 99.44% and molecular weight is 304.30 g/mol.

  • Water-soluble fluorescent probe for spectroscopic assays.
  • High purity suitable for analytical and fluorimetric analysis.
  • Solid powder form for convenient storage and handling.
  • Molecular weight 304.30 g/mol for accurate formulation.
  • CAS number 59323-54-5 for unambiguous identification.

Catalog No. 50-002-82656 Supplier Medchemexpress LLC Supplier No. HYW127714250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Pyrenesulfonic acid sodium is the sodium salt of 1-pyrenesulfonic acid, a water-soluble, fluorescent aromatic sulfonate supplied as a solid research reagent. It is used as a fluorescent probe and analytical standard in life-science and chemical research; typical listed purity is 99.44% and molecular weight is 304.30 g/mol.

  • Water-soluble fluorescent probe for spectroscopic assays.
  • High purity suitable for analytical and fluorimetric analysis.
  • Solid powder form for convenient storage and handling.
  • Molecular weight 304.30 g/mol for accurate formulation.
  • CAS number 59323-54-5 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.