Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(pyrimidin-5-yl)ethan-1-ol | 79691-74-0 | 96.6% | 124.14 g·mol⁻¹ | C6H8N2O | 250 MG
SDP

Catalog No. 5000209658
Encompass_Preferred

1-(Pyrimidin-5-yl)ethan-1-ol is a pyrimidine-derived small-molecule intermediate used in organic and medicinal chemistry for the synthesis of pyrimidine-containing active compounds. It is supplied as a research-use reagent with documented purity and analytical certification.

  • Used as an intermediate in organic and medicinal chemistry syntheses.
  • Molecular formula C6H8N2O and molecular weight 124.14 g·mol⁻¹.
  • Purity typically 96.6% by HPLC (certificate of analysis available).
  • Provided in small milligram-scale quantities suitable for research applications.
  • Stored under recommended conditions to preserve stability.

Catalog No. 50-002-09658 Supplier Medchemexpress LLC Supplier No. 250MGHYW062571
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(Pyrimidin-5-yl)ethan-1-ol is a pyrimidine-derived small-molecule intermediate used in organic and medicinal chemistry for the synthesis of pyrimidine-containing active compounds. It is supplied as a research-use reagent with documented purity and analytical certification.

  • Used as an intermediate in organic and medicinal chemistry syntheses.
  • Molecular formula C6H8N2O and molecular weight 124.14 g·mol⁻¹.
  • Purity typically 96.6% by HPLC (certificate of analysis available).
  • Provided in small milligram-scale quantities suitable for research applications.
  • Stored under recommended conditions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.