Learn More
Medchemexpress LLC 1-pyrrolidine-2,2,3,3,4,4-d6-acetamide, α-ethyl-5-oxo-, (αS)- | 1133229-30-7 | MFCD08063556 | 98.7% | 176.25 g/mol | C8H8D6N2O2 | 5 MG

Supplier: Medchemexpress LLC HY1102255MG
Levetiracetam-d6 (ring-d6) is a deuterated analog of levetiracetam used as an internal standard and tracer for quantitative analysis of levetiracetam in biological matrices. It supports accurate isotope-dilution measurements and is provided with analytical documentation for laboratory assays.
- High deuterium incorporation to minimize isotopic interference.
- Suitable for LC-MS, GC-MS, and NMR quantitative assays.
- Purity 98.7% as determined by LC-MS.
- Molecular weight 176.25 g/mol and formula C8H8D6N2O2 for unambiguous identification.
- Supplied with certificate of analysis and product data sheet.
- Intended for use as an internal standard in biological sample analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.