Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pyrrolidineacetonitrile, 97%, Thermo Scientific Chemicals

Catalog No. AAL0173706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0173706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0173706 Supplier Thermo Scientific Chemicals Supplier No. L0173706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 29134-29-0 | C6H10N2 | 110.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29134-29-0
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00014098
InChI Key NPRYXVXVLCYBNS-UHFFFAOYSA-N
Synonym 1-pyrrolidineacetonitrile, 1-pyrrolidino acetonitrile, 2-pyrrolidin-1-yl acetonitrile, pyrrolidine-1-acetonitrile, pyrrolidin-1-ylacetonitrile, pyrrolidinoacetonitrile, 1-cyanomethylpyrrolidine, n-pyrrolidinoacetonitrile, unii-6p9eel10k2, pyrroldinoacetonitrile
PubChem CID 95270
IUPAC Name 2-pyrrolidin-1-ylacetonitrile
SMILES C1CCN(C1)CC#N

Specifications

Refractive Index 1.4563
Quantity 5 g
UN Number UN3276
Beilstein 106157
Formula Weight 110.16
Percent Purity 97%
Chemical Name or Material 1-Pyrrolidineacetonitrile
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 249-463-2
RTECSNumber UX9678600
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.