Learn More
Medchemexpress LLC 1-Pyrrolidinecarbodithioic acid, ammonium salt (1:1) | 5108-96-3 | 98.0% | 164.29 | 10 G
SDP
Catalog No. 5000387869
Encompass_Preferred
Pyrrolidinedithiocarbamate ammonium is a selective and blood-brain barrier (BBB) permeable NF-κB inhibitor. It is useful for the treatment of ulcerative colitis by suppressing inflammatory cytokines.
  • Attenuates IL-8 production
  • Suppresses NF-κB activation
  • Inhibits NF-κB DNA binding and transcriptional activity
  • Reduces intestinal length shortening
  • Decreases DAI score
  • Lowers IL-1β and TNF-α levels
Catalog No. 50-003-87869 Supplier Medchemexpress LLC Supplier No. HY1873810G
Add to Cart
Add to Cart
Pyrrolidinedithiocarbamate ammonium is a selective and blood-brain barrier (BBB) permeable NF-κB inhibitor. It is useful for the treatment of ulcerative colitis by suppressing inflammatory cytokines.
  • Attenuates IL-8 production
  • Suppresses NF-κB activation
  • Inhibits NF-κB DNA binding and transcriptional activity
  • Reduces intestinal length shortening
  • Decreases DAI score
  • Lowers IL-1β and TNF-α levels