Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-pyrrolidinecarboxylic acid, 2-ethenyl-, 1,1-dimethylethyl ester (2S) | 115393-77-6 | MFCD22381778 | 97.9% | 197.27 | C11H19NO2 | 5 G
SDP

Catalog No. 5000210704
Encompass_Preferred

tert-Butyl (S)-2-vinylpyrrolidine-1-carboxylate is a chiral small-molecule drug intermediate used in the synthesis of active pharmaceutical compounds. It is supplied in laboratory-scale quantities and is typically handled and stored under an inert atmosphere to preserve integrity.

  • Chiral vinylpyrrolidine intermediate for synthetic applications.
  • CAS number 115393-77-6.
  • Molecular formula C11H19NO2; molecular weight 197.27.
  • Available in small-scale packages (100 mg-50 g).
  • Store under inert atmosphere at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Batch COAs report purity values around 97.9% (GC); some batches report 99.2% - check COA for the specific lot.

Catalog No. 50-002-10704 Supplier Medchemexpress LLC Supplier No. HYW066517A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl (S)-2-vinylpyrrolidine-1-carboxylate is a chiral small-molecule drug intermediate used in the synthesis of active pharmaceutical compounds. It is supplied in laboratory-scale quantities and is typically handled and stored under an inert atmosphere to preserve integrity.

  • Chiral vinylpyrrolidine intermediate for synthetic applications.
  • CAS number 115393-77-6.
  • Molecular formula C11H19NO2; molecular weight 197.27.
  • Available in small-scale packages (100 mg-50 g).
  • Store under inert atmosphere at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Batch COAs report purity values around 97.9% (GC); some batches report 99.2% - check COA for the specific lot.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.