Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Pyrrolidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, (3S)- | 147081-44-5 | 99.95% | 186.25 | 10 G
SDP

Catalog No. 5000404546
Encompass_Preferred

(S)-1-Boc-3-aminopyrrolidine is a biochemical reagent suitable for use as a biological material or organic compound in life science research. It is intended for research use only and not for sale to patients.

  • Purity: 99.95%
  • Molecular weight: 186.25
  • Formula: C9H18N2O2
  • CAS No.: 147081-44-5
  • Appearance: Liquid (density: 1.067 g/cm3)
  • Color: Colorless to light yellow
  • Solubility (in H2O): 100 mg/mL (536.91 mM; requires ultrasonic)

Catalog No. 50-004-04546 Supplier Medchemexpress LLC Supplier No. HY4009810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-1-Boc-3-aminopyrrolidine is a biochemical reagent suitable for use as a biological material or organic compound in life science research. It is intended for research use only and not for sale to patients.

  • Purity: 99.95%
  • Molecular weight: 186.25
  • Formula: C9H18N2O2
  • CAS No.: 147081-44-5
  • Appearance: Liquid (density: 1.067 g/cm3)
  • Color: Colorless to light yellow
  • Solubility (in H2O): 100 mg/mL (536.91 mM; requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.