Learn More
Cayman Chemical 1 R TrIfluoromethyloleyl a
SDP
Catalog No. 502949533
Encompass_Preferred
An oleic acid analog; inhibits erastin-induced ferroptosis in primary fibroblasts isolated from patients with Friedreich ataxia at 5, 10, or 20 µM; reduces FXN siRNA knockdown-induced lipid peroxidation in NBT human myoblasts at 5 µM
Catalog No. 50-294-9533 Supplier Cayman Chemical Supplier No. 31796500 UG
An oleic acid analog; inhibits erastin-induced ferroptosis in primary fibroblasts isolated from patients with Friedreich ataxia at 5, 10, or 20 µM; reduces FXN siRNA knockdown-induced lipid peroxidation in NBT human myoblasts at 5 µM