Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-1-{(Rp)-2-[2-(Diphenylphosphino)Phenyl]Ferrocenyl}Ethylbis[3,5-Bis-(Trifluoromethyl)Phenyl]Phosphine

Catalog No. 65671100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
5 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-671-100G 100 g
65-671-50G 50 g
65-671-5G 5 g
65-671-500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 65-671-100G Supplier Sigma Aldrich Supplier No. 65671100G
Only null left
Add to Cart
Add to Cart
Request a Quote

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The product is an air-stable and non-hygroscopic Solvias chiral phosphine ligand.Walphos SL-W001-1, in the presence of a ruthenium catalyst may be used as a ligand in the following processes: Asymmetric hydrogenation of IPCA-D, a cinnamic acid derivative.Addition of 3,5-difluorophenylketimine to 4-methoxyphenylboronic acid to form the corresponding tertiary substituted cyclic sulfamate derivative with high enantioselctivity.2sold in collaboration with Solvias AG

Specifications

CAS 387868-06-6
Molecular Formula C46H32F12FeP2
Linear Formula C46H32F12FeP2
MDL Number MFCD08561122
Quantity 100 g
Synonym (1S)-1-[(1R)-1-[Bis[3,5-bis(trifluoromethyl)phenyl]phosphino]ethyl]-2-[2-(diphenylphosphino)phenyl]ferrocene (acc to CAS); Walphos SL-W001-1
Molecular Weight (g/mol) 930.52 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.