Learn More
New England Biolabs, Inc. O-Glycosidase and Neuraminidase Bundle – 1 bundle
SDP
Supplier:  New England Biolabs, Inc. E0540S
Encompass
This bundle contains 1 vial each of O-Glycosidase and a2-3,6,8 Neuraminidase, which can be used simultaneously for the removal of terminal sialic acid residues and core 1 and core 3 O-glycans.
Catalog No. NC0440311