Learn More
Fritsch Milling And Sizing Inc 1 Set Sample Glasses 12X Glasses with Lids
SDP
Catalog No. 501534708
Encompass
Set = 12 pieces of sample glasses 1.5 ml volume with lids for grinding bowl made of stainless steel for single-use technology for PULVERISETTE 7 premium line.
Catalog No. 50-153-4708 Supplier Fritsch Milling And Sizing Inc Supplier No. 83.3165.00
Set = 12 pieces of sample glasses 1.5 ml volume with lids for grinding bowl made of stainless steel for single-use technology for PULVERISETTE 7 premium line.