Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-1-[(Sp)-2-(Dicyclohexylphosphino)Ferrocenyl]Ethyldi-Tert-Butylphosphine

Catalog No. 887331G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-733-1G 1 g
88-733-100MG 100 mg
88-733-500MG 500 mg
88-733-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 88-733-1G Supplier Sigma Aldrich Supplier No. 887331G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

sold in collaboration with Solvias AGIt may be used as a ligand in the palladium catalyzed hetero cross-coupling between aryl bromides/aryl triflates with potassium thioacetate to form S-aryl thioacetates.

Specifications

CAS 158923-11-6
Molecular Formula C32H52FeP2
Linear Formula C32H52FeP2
MDL Number MFCD08543435
Quantity 1 g
Synonym (2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(dicyclohexylphosphino)ferrocene (acc to CAS); Josiphos SL-J009-1
Molecular Weight (g/mol) 554.55 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.