Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-1-[(Sp)-2-(Dicyclohexylphosphino)Ferrocenylethyl]Diphenylphosphine

Catalog No. 887231G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-723-1G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-723-1G Supplier Sigma Aldrich Supplier No. 887231G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

sold in collaboration with Solvias AG™-1-[(SP)-2-(Dicyclohexylphosphino)ferrocenylethyl]diphenylphosphine can be used as a ligand:To prepare iridium metal complex (IrIJOSIPHOS), which is used as a catalyst in the asymmetric hydrogenation of 2-phenylquinoxaline.In the preparation of natural products by asymmetric conjugate addition of Grignard reagents to +a-substituted cyclic alkenones.

Specifications

CAS 167416-28-6
Molecular Formula C36H56FeP2
Linear Formula C36H56FeP2
MDL Number MFCD01074441
Quantity 100 mg
Synonym (1R)-1-(Dicyclohexylphosphino)-2-[(1R)-1-(dicyclohexylphosphino)ethyl]ferrocene (acc to CAS); Josiphos SL-J003-1
Molecular Weight (g/mol) 606.62 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.