Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-1-[(Sp)-2-(Diphenylphosphino)Ferrocenyl]Ethyldicyclohexylphosphine

Catalog No. 887175G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
50 g
100 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-717-5G 5 g
88-717-100G 100 g
88-717-50G 50 g
88-717-100MG 100 mg
88-717-500MG 500 mg
88-717-1G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 88-717-5G Supplier Sigma Aldrich Supplier No. 887175G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

sold in collaboration with Solvias AGRhodium complex of ™-(S)-JOSIPHOS can be used as a catalyst for the selective asymmetric hydrogenation of: Z-+a-aryl-+a-(enamido)phosphonateso-alkoxy tetrasubstituted enamides to yield +a-amino alcohol analogs with excellent enantiomeric excess.

Specifications

CAS 162291-02-3
Molecular Formula C36H44FeP2 -+ C2H5OH
Linear Formula C36H44FeP2 -+ C2H5OH
MDL Number MFCD00800285
Quantity 5 g
Synonym (2S)-1-[(1S)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene (acc to CAS); (S)-(R)-Josiphos; Josiphos SL-J001-2
Molecular Weight (g/mol) 640.6 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.