Learn More
Enzo Life Sciences D-erythro-Sphingosine 1-phosphate (caged) (500µg)
SDP
Supplier:  Enzo Life Sciences ALX306017C500
Encompass_Preferred
A photolyzable derivative of sphingosine 1-phosphate (1-SPP) (BML-SL140). Important tool for the study of 1-SPP mediated intracellular events. Alternative name: 1-SPP (caged). Purity: ≥95%. Appearance: Clear to slight yellow oil. Solubility: Soluble in methanol or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8661