Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 1-Step Human High-Yield IVT Kit

Catalog No. p-4532178 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 100 μL
10 x 100 μL
2 x 2 mL
Type:
1-step Human High-yield Maxi IVT Kit
1-step Human High-yield Mini IVT Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
PI88890 2 x 100 μL 1-step Human High-yield Mini IVT Kit
PI88891 10 x 100 μL 1-step Human High-yield Mini IVT Kit
PI88892 2 x 2 mL 1-step Human High-yield Maxi IVT Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI88890 Supplier Thermo Scientific™ Supplier No. 88890
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Express functional proteins with 10 to 100 times greater yield per mL than other mammalian IVT systems with Thermo Scientific™ 1-Step Human High-Yield In Vitro Translation (IVT) Kits.

The Thermo Scientific™ 1-Step Human High-Yield In Vitro Translation (IVT) Kits with continuous-feed device enables the expression of functional proteins with 10 to 100 times greater yield per mL than other eukaryotic IVT translation systems.

The 1-Step High-Yield IVT System uses modified HeLa cell extracts to take advantage of the robust human transcription and translation machinery and generate functional full-length proteins. In this system, protein expression is performed in a proprietary mini dialysis device that allows a continuous supply of nucleotides, amino acids and energy generating substrates into the reaction while removing inhibitors of proteins synthesis. This continuous-exchange cell-free (CECF) system enables protein expression for up to 24 hours and produces protein yields ranging from 250 to 750μg/mL. The complete kits come with all components required for proper transcription and translation of recombinant protein, including an optimized expression vector.

Highlights:

  • High expression – up to 750μg/mL of expressed protein

  • Reproducibility – low variability between experiments

  • Fast – express high levels of protein with an overnight incubation

  • Functional – obtain more active protein than with bacterial protein expression systems

1-Step Protein Expression:

  • Streamlined reaction – requires only five components

  • Easy-to-use – transcription and translation is performed in one reaction step

  • Adapt for labeling – amenable to incorporation of heavy or unnatural amino acids

Includes:

Complete kits contain cell lysate, reaction mix, accessory proteins, 5X dialysis buffer, control DNA, cloning vector, mini dialysis devices and centrifuge tubes

Requires:

30°C incubator or water bath

Recommended for:

Production of recombinant proteins for structural analysis; Generate active proteins for protein interaction studies; Perform rapid expression and characterization of mutant proteins; Small-scale production of virual particles; Incorporation of unnatural amino acids or toxic protein production

TRUSTED_SUSTAINABILITY

Specifications

Type 1-step Human High-yield Mini IVT Kit
Sufficient For 2 Reactions of 100 μL Each
Quantity 2 x 100 μL
Content And Storage Sufficient For: 2 reactions of 100 μL each
• HeLa Lysate, 110 μL
• Accessory Proteins, 25 μL
• Reaction Mix, 40 μL
• 5X Dialysis Buffer, 600 μL
• Positive Control DNA (pCFE-GFP), 10 μg
• pT7CFE1-NHis-GST-CHA, 10 μg
• Microdialysis Device, 2 each
• Nuclease-free Water, 5 mL

Store micro-dialysis devices and nuclease-free water at room temperature. Store all other components at -80°C.
Expression System Cell-Free
Expressed Protein Type Insoluble Protein
Product Line 1-Step
When comparing 1-Step Human High-Yield Mini IVT Kit with 1-Step Human Coupled IVT Kit - DNA, is one faster than the other?

No, both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA can be run overnight (depends upon the gene).

Do both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA provide the same purity of protein?

Yes, both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA provide the same purity of protein.

I'm using the 1-Step Human High-Yield Mini IVT Kit. Can I get the vector map for the pT7CFE1-NHis-GST-CHA Vector from the kit?

The pT7CFE1-NHis-GST-CHA vector is also offered as a standalone product (Cat. No. 88871) and the vector information can be accessed on the product page (https://www.thermofisher.com/order/catalog/product/88871), under "Vector information" at the bottom of the page.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.